Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Myoglobin Rabbit Polyclonal Antibody with BSA and azide

Catalog No. RB9068R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB9068R1 1 mL Prediluted
RB9068P1 500 μL Concentrated
RB9068R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RB9068R1 Supplier Epredia™ Supplier No. RB9068R1

Epredia™ Myoglobin, Rabbit Polyclonal Antibody provides accurate, reproducible results.

Myoglobin is a cytoplasmic, single chain polypeptide of 153 amino acids and contains a single heme group. Myoglobin is found in skeletal and carciac muscle but not in smooth muscle and functions as an oxygen transporting pigment. Upon damage to the muscle cell due to infarction of a coronary artery, neurological trauma, infection of tumor processes, myoglobin escapes to the environment and can be found in plasma using sensitive assays.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: Purified human myoglobin

Molecular Weight: 17.8kDa

Positive Control: Skeletal muscle

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Myoglobin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Immunogen Purified human myoglobin
Quantity 1 mL
Regulatory Status RUO
Research Discipline Muscle Biology
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.