Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Myosin, Smooth Muscle Heavy Chain, Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS1177RQ Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
Prediluted
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1177RQ 12 mL Ready-to-Use Prediluted
MS1177P0 100 μL Concentrated 200μg/mL
MS1177P1ABX 100 μL Concentrated 1mg/mL
MS1177PABX 200 μL Concentrated 1mg/mL
MS1177P1 500 μL Concentrated 200μg/mL
MS1177R1 1 mL Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS1177RQ Supplier Epredia™ Supplier No. MS1177RQ

Provides accurate, reproducible results with the Epredia™ Myosin, Smooth Muscle Heavy Chain Ab-1, Mouse Monoclonal Antibody.

SM-MHC is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmently regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2.

Host Species: Mouse

Clone: SMMS-1

Isotype: IgG1/k

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Crude human uterus extract

Molecular Weight: 205kDa (MHC-1) and 200kDa (MHC-2)

Positive Control: Uterus

Cellular Localization: Cytoplasmic

Recommended for:

Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Myosin, Smooth Muscle Heavy Chain Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SMMS-1
Concentration Prediluted
Conjugate Unconjugated
Host Species Mouse
Immunogen Crude human uterus extract
Quantity 12 mL
Regulatory Status IVD
Research Discipline Muscle Biology
Primary or Secondary Primary
Form Ready-to-Use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.