Learn More
Epredia™ Lab Vision™ Negative Control for Mouse IgG2b Ab-1 with BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: NCG2B.01
Isotype: IgG2b/κ
Species Reactivity: All species.
Recommended for:
Negative control for:
- Immunohistochemistry (Frozen and formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation
- Flow Cytometry
- ELISA
- Gel Supershift
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Negative Control for Mouse IgG2b Ab-1 |
| Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | NCG2B.01 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Research Discipline | Negative Controls |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.