Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Neurofilament (200kDa & 68kDa) Ab-1, Mouse Monoclonal Antibody

Catalog No. MS359S1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS359S1 500 μL Concentrated
MS359S0 100 μL Concentrated
MS359R7 7 mL Prediluted
MS359R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS359S1 Supplier Epredia™ Supplier No. MS359S1
Add to Cart
Add to Cart

Take advantage of a quality product that delivers accurate results with the Epredia™ Neurofilament (200kDa & 68kDa) Ab-1, Mouse Monoclonal Antibody.

Neurofilaments are the intermediate filaments of neurons and their processes. They are protein triplets composed of three major subunits of MW 68kDa, 155kDa, and 200kDa. Neurofilaments are expressed in tumors of neural origin, or displaying neuronal differentiation, such as neuroblastoma, medulloblastoma, and retinoblastoma.

Host Species: Mouse

Clone: 2F11

Isotype: IgG1/κ

Species Reactivity: Human, Rabbit, Cat, Mouse and Rat. Does not react with dog. Others not known.

Epitope: Not determined

Immunogen: Neurofilament purified from human brain

Molecular Weight: 68 and 200kDa (phosphorylated forms of thsese subunits)

Positive Control: IMR-5 cells, Brain

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Specifications

Antigen Neurofilament (200kDa and 68kDa) Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2F11
Conjugate Unconjugated
Host Species Mouse
Immunogen Neurofilament purified from human brain
Quantity 500 μL
Regulatory Status IVD
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Feline, Human, Mouse, Rabbit, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.