Learn More
Neurotrophin Receptor/NGF-Receptor (p75NGFR) Ab-1 Mouse Monoclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: NGFR5
Isotype: IgG1/κ
Species Reactivity: Human, Monkey (Cebus apella, Macaca fascicularis, mulatta, and nemestrina), Baboon (Papio anubis and Cynocephalus), Cat, Rabbit, Ferret. Does not react with mouse and rat.
Epitope: aa 1-160
Immunogen: NGFR from A875 melanoma cells
Molecular Weight: 75kDa
Positive Control: Melanoma
Cellular Localization: Cell surface and cytoplasmic
Recommended for:
- Immunohistochemistry (Not suitable)
- Western Blotting (Not suitable)
- Immunofluorescence
- Flow Cytometry
- Inhibits Binding of NGF to Melanoma Cells
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Neurotrophin Receptor/NGF-Receptor (p75superscriptNGFRsuperscript) Ab-1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | NGFR5 |
| Concentration | 200μg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Immunogen | NGFR from A875 melanoma cells |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.