Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurotrophin Receptor/NGF-Receptor (p75NGFR) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS394P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
MS394P 1 mL Unconjugated 200μg/mL
MS394B1 500 μL Biotin 200μg/mL
MS394B 1 mL Biotin 200μg/mL
MS394B0 100 μL Biotin 200μg/mL
MS394P0 100 μL Unconjugated 200μg/mL
MS394P1 500 μL Unconjugated 200μg/mL
MS394P1ABX 100 μL Unconjugated 1mg/mL
MS394PABX 200 μL Unconjugated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS394P Supplier Epredia Supplier No. MS394P

Epredia™ Neurotrophin Receptor/NGF-Receptor (p75NGFR) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

NGF is important for the development, differentiation, and survival of variety of neuronal and non-neuronal cells. Its action is mediated by binding two distinct receptors, the high affinity p140 and low affinity Nerve Growth Factor (NGF) Receptor (p75NGFR) or Neurotrophin Receptor (p75NTR).

Host Species: Mouse

Clone: NGFR5

Isotype: IgG1/κ

Species Reactivity: Human, Monkey (Cebus apella, Macaca fascicularis, mulatta, and nemestrina), Baboon (Papio anubis and Cynocephalus), Cat, Rabbit, Ferret. Does not react with mouse and rat.

Epitope: aa 1-160

Immunogen: NGFR from A875 melanoma cells

Molecular Weight: 75kDa

Positive Control: Melanoma

Cellular Localization: Cell surface and cytoplasmic

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting (Not suitable)
  • Immunofluorescence
  • Flow Cytometry
  • Inhibits Binding of NGF to Melanoma Cells

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Neurotrophin Receptor/NGF-Receptor (p75superscriptNGFRsuperscript) Ab-1
Applications Flow Cytometry
Classification Monoclonal
Clone NGFR5
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen NGFR from A875 melanoma cells
Quantity 1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Feline, Ferret, Human, Non-human Primate, Rabbit
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.