Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ NFκB/p65 (Rel A) Rabbit Polyclonal Antibody with BSA and azide

Catalog No. RB9034R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
Form:
Prediluted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB9034R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RB9034R1 Supplier Epredia™ Supplier No. RB9034R1

Take advantage of a quality product that delivers accurate results with the Epredia™ NFκB/p65 (Rel A), Rabbit Polyclonal Antibody.

NFκB is a heterodimer consisting of a 50kDa DNA binding subunit and a 65kDa transactivating subunit (RelA). The p50/p65 heterodimer is present in the cytoplasm in inactive form bound to its inhibitor, IκB. In unstimulated cells, nuclear localization signals present on p65 are masked by members of the IκB family of inhibitory proteins. Subsequent to cell stimulation, IκB undergoes phosphorylation, ubiquitination and degradation by a proteosome-dependent pathway, allowing nuclear translocation of the active dimeric NFκB transcription factor.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Rat and Rabbit. Others not tested.

Epitope: Internal domain

Immunogen: A synthetic peptide corresponding to internal domain of human NFκB/p65

Molecular Weight: 65kDa

Positive Control: Prostate gland and placenta

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen NFkappaB/p65 (Rel A)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal domain of human NFkappaB p65
Quantity 1 mL
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Target Species Human, Rabbit
Form Prediluted
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.