Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Nitric Oxide Synthase with BSA and azide

Take advantage of a quality product that delivers accurate results with the Epredia™ Nitric Oxide Synthase, endothelial (eNOS), Rabbit Polyclonal Antibody.

Supplier:  Epredia™ RB9279P0

 View more versions of this product

Catalog No. RB9279P0


Description

Description

NOS oxidizes a guanidine nitrogen of arginine releasing nitric oxide in the form of a free radical and citrulline. Nitric oxide thus generated acts as a messenger in diverse functions including vasodilation neurotransmission, anti-tumor and anti-pathogenic activities. NOS is classified under three types: neuronal NOS(nNOS) or brain NOS (bNOS); induciblr NOS (iNOS) or macrophage NOS (mNOS); and endothelial NOS (eNOS). This antibody reacts with eNOS.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat, Dog, Pig and Cow. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from C-ternimal of human eNOS

Molecular Weight: 135kDa

Positive Control: Capillary endothelium, regenerating endothelium

Cellular Localization: Cytoplasmic This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Nitric Oxide Synthase, endothelial (eNOS)
Polyclonal
Unconjugated
A synthetic peptide derived from C-ternimal of human eNOS
RUO
Primary
Concentrated
Immunohistochemistry (Paraffin)
200µg/mL
Rabbit
100 μL
Neuroscience
Canine, Human, Mouse, Rat
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.