Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Nitric Oxide Synthase with BSA and azide

Take advantage of a quality product that delivers accurate results with the Epredia™ Nitric Oxide Synthase, inducible (iNOS), Rabbit Polyclonal Antibody.

Specifications

Antigen Nitric Oxide Synthase, inducible (iNOS)
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
RB9242R1
View Documents
Epredia™
RB9242R1
Prediluted 1 mL N/A N/A N/A
Description

Description

NOS oxidizes a guanidine nitrogen of arginine releasing nitric oxide in the form of a free radical and citrulline. Nitric oxide thus generated acts as a messenger in diverse functions including vasodilation neurotransmission, anti-tumor and anti-pathogenic activities. Inducible NOS (iNOS) or macrophage NOS (mNOS) is a calcium/calmodulin independent and is expressed in activated macrophages and stimulated glial cells.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Rat, Mouse and Human. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide derived from C-ternimal of mouse iNOS

Molecular Weight: 130kDa

Positive Control: Lung

Cellular Localization: Cytoplasmic This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Nitric Oxide Synthase, inducible (iNOS)
Polyclonal
Rabbit
Neuroscience
IgG
Immunohistochemistry (Paraffin)
Unconjugated
RUO
A synthetic peptide derived from C-ternimal of mouse iNOS
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.