Learn More
Description
Host Species: Mouse
Clone: NY-BR-1No. 2
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: a 77 aa peptide from human NYBR-1 protein
Molecular Weight: 150-160kDa
Positive Control: Breast carcinoma
Cellular Localization: Predominantly cytoplasmic and occasional nuclear
Recommended for:
Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | NY-BR-1 |
| Applications | Immunohistochemistry |
| Clone | NY-BR-1 No.2 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | a 77 aa peptide from human NYBR-1 protein |
| Quantity | 1 mL |
| Primary or Secondary | Primary |
| Target Species | Human |
| Form | Ready-to-Use |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.