Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ NY-BR-1 Mouse Monoclonal Antibody

Catalog No. MS1932RQ Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
7 mL
12 mL
Form:
Purified
Ready-to-Use
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1932RQ 12 mL Ready-to-Use
MS1932P 1 mL Purified
MS1932P0 100 μL Purified
MS1932R1 1 mL Ready-to-Use
MS1932R7 7 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1932RQ Supplier Epredia™ Supplier No. MS1932RQ

Take advantage of a quality product that delivers accurate results with the Epredia™ NY-BR-1, Mouse Monoclonal Antibody.

NY-BR-1 is a novel differentiation marker predominantly expressed in normal breast and breast cancer. It has been shown by Immunohistochemistry (IHC) that NY-BR-1 was present solely in ductal epithelium of normal breast tissue. Invasive carcinoma of the breast and carcinoma in situ were positive for NY-BR-1, whereas most other tumors and normal tissues are negative, with the exceptions of one-third of sweat gland carcinomas and around two percent of prostate carcinoma. NY-BR-1 is reported to express more frequently in estrogen-positive and lymph node-negative primary carcinomas, and more commonly in grade 1 than in grade 2 or grade 3. In a study by Varga et al (2006) forty-nine percent of lymph node metastasis were shown to be positive.

Host Species: Mouse

Clone: NY-BR-1No. 2

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: a 77 aa peptide from human NYBR-1 protein

Molecular Weight: 150-160kDa

Positive Control: Breast carcinoma

Cellular Localization: Predominantly cytoplasmic and occasional nuclear

Recommended for:

Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen NY-BR-1
Applications Immunohistochemistry
Clone NY-BR-1 No.2
Conjugate Unconjugated
Host Species Mouse
Immunogen a 77 aa peptide from human NYBR-1 protein
Quantity 12 mL
Primary or Secondary Primary
Form Ready-to-Use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.