Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ p130/Rb2 Ab-2 Mouse Monoclonal Antibody with BSA and azide

Take advantage of a quality product that delivers accurate results with the Epredia™ p130/Rb2 Ab-2, Mouse Monoclonal Antibody.

Supplier:  Epredia™ MS866R1

 View more versions of this product

Catalog No. MS866R1


Description

Description

Two retinoblastoma-related proteins, p107 and pRb2/p130 are structurally and functionally similar to the product of the retinoblastoma gene (pRb/p105). Like pRb they bind transforming proteins of DNA tumor viruses through a particular region called the “pocket domain”. Both proteins play a fundamental role in growth control. The highest percentage of undetectable levels and the tightest inverse correlation with the histological grading and with PCNA expression in the most aggressive tumor types were found for pRb2/p130, which may suggest an important role for this protein in the pathogenesis and progression of lung cancer.

Host Species: Mouse

Clone: 130P215; same as DCS-215

Isotype: IgG1

Species Reactivity: Human and Rat. Others not known.

Epitope: aa 878-913

Immunogen: A synthetic peptide from p130 corresponding to aa 878-913

Molecular Weight: 130kDa

Positive Control: A427 cells, Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

p130/Rb2 Ab-2
Monoclonal
Unconjugated
Mouse
1 mL
Cell Cycle
Human, Rat
IgG1
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
130P215; same as DCS-215
Ready-to-use for IHC (UVLP)
A synthetic peptide from p130 corresponding to aa878-913
RUO
Primary
Prediluted
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.