Learn More
Description
Host Species: Mouse
Clone: F4
Isotype: IgG1
Species Reactivity: Human, Chinese Hamster. Does not react with mouse. Others not known.
Epitope: Extracellular amino terminal half. This epitope is resistant to formalin fixation and periodate oxidation and is different from those of other commonly known MAbs to p-glycoprotein, such as C219, JSB-1, and MRK16, which recognize an intracellular and extracellular epitope of p-glycoprotein, respectively.
Immunogen: Human and hamster drug-resistant viable cells followed by crude plasma membranes
Molecular Weight: 170kDa
Positive Control: Drug-resistant cells, Kidney or breast CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Flow Cytometry (Fixed cells only)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | p170/p-Glycoprotein/MDR Ab-2 |
| Applications | Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | F4 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human and hamster drug-resistant viable cells followed by crude plasma membranes |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.