Learn More
Description
Host Species: Mouse
Clone: 18P118; same as DCS-118
Isotype: IgG1
Species Reactivity: Human, Mouse. Others not known.
Epitope: Not determined
Immunogen: Purified recombinant p18INK4c protein
Molecular Weight: 18kDa
Positive Control: HeLa cells; Tonsil, skeletal muscle, heart, pancreas
Cellular Localization: Cytoplasmic and Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | p18superscriptINK4csuperscript Ab-3 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 18P118; same as DCS-118 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Purified recombinant p18superscriptINK4csuperscript protein |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.