Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p18INK4c Ab-3 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS858P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS858P1 500 μL Concentrated 200μg/mL
MS858P0 100 μL Concentrated 200μg/mL
MS858P1ABX 100 μL Concentrated 1mg/mL
MS858PABX 200 μL Concentrated 1mg/mL
MS858P 1 mL Concentrated 200μg/mL
MS858R1 1 mL Prediluted -
MS858R7 7 mL Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS858P1 Supplier Epredia Supplier No. MS858P1

Ensure accurate, reproducible results with the Epredia™ p18INK4c Ab-3, Mouse Monoclonal Antibody.

p18INK4c is a specific inhibitor of cdk4/cdk6 and is expressed during G1 -to S transition in the eukaryotic cell division cycle. It is maximally induced as cells enter S-phase.

Host Species: Mouse

Clone: 18P118; same as DCS-118

Isotype: IgG1

Species Reactivity: Human, Mouse. Others not known.

Epitope: Not determined

Immunogen: Purified recombinant p18INK4c protein

Molecular Weight: 18kDa

Positive Control: HeLa cells; Tonsil, skeletal muscle, heart, pancreas

Cellular Localization: Cytoplasmic and Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen p18superscriptINK4csuperscript Ab-3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 18P118; same as DCS-118
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified recombinant p18superscriptINK4csuperscript protein
Quantity 500 μL
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Target Species Human, Mouse
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.