Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p21 WAF1 Ab-11 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS891B1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
MS891B1 500 μL Biotin 200μg/mL
MS891P0 100 μL Unconjugated 200μg/mL
MS891PABX 200 μL Unconjugated 1mg/mL
MS891B0 100 μL Biotin 200μg/mL
MS891B 1 mL Biotin 200μg/mL
MS891P1 500 μL Unconjugated 200μg/mL
MS891P 1 mL Unconjugated 200μg/mL
MS891P1ABX 100 μL Unconjugated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS891B1 Supplier Epredia Supplier No. MS891B1

Ensure accurate, reproducible results with the Epredia™ p21WAF1 Ab-11, Mouse Monoclonal Antibody.

p21WAF1/Cip1/Sdi1/Pic1 is a tumor suppressor protein. Expression of p21WAF1 is induced by wild type, but not mutant, p53 suppressor protein. The p21WAF1 protein binds to cyclin/CDK complexes and inhibits their kinase activity thereby stopping cell cycle progression. It also binds to PCNA (proliferating cell nuclear antigen) and blocks DNA replication but not the DNA repair process.

Host Species: Mouse

Clone: CP74

Isotype: IgG2b

Species Reactivity: Human and Rat. Others not known.

Epitope: aa 1-80

Immunogen: Full-length human recombinant p21 protein

Molecular Weight: 21kDa

Positive Control: Raji, PC12 cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native verified)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen p21superscriptWAF1superscript Ab-11
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone CP74
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen Full length human recombinant p21 protein
Quantity 500 μL
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Target Species Human, Rat
Form Concentrated
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.