Learn More
Description
Host Species: Mouse
Clone: CP74
Isotype: IgG2b
Species Reactivity: Human and Rat. Others not known.
Epitope: aa 1-80
Immunogen: Full-length human recombinant p21 protein
Molecular Weight: 21kDa
Positive Control: Raji, PC12 cells
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Native verified)
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | p21superscriptWAF1superscript Ab-11 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | CP74 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Full length human recombinant p21 protein |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.