Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ p21WAF1 Ab-5 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS387R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
Form:
Prediluted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS387R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS387R1 Supplier Epredia™ Supplier No. MS387R1

Achieve accurate, reproducible results with the Epredia™ p21WAF1 Ab-5, Mouse Monoclonal Antibody.

p21WAF1/Cip1/Sdi1/Pic1 is a tumor suppressor protein. Expression of p21WAF1 is induced by wild type, but not mutant, p53 suppressor protein. The p21WAF1 protein binds to cyclin/CDK complexes and inhibits their kinase activity thereby stopping cell cycle progression. It also binds to PCNA (proliferating cell nuclear antigen) and blocks DNA replication but not the DNA repair process.

Host Species: Mouse

Clone: HZ52

Isotype: IgG1

Species Reactivity: Human, Cow, Mouse, and Rat. Others not known.

Epitope: Not determined

Immunogen: Full-length human recombinant p21 protein

Molecular Weight: 21kDa

Positive Control: Ls174T or Raji cells, Colon CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunoprecipitation
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen p21superscriptWAF1superscript Ab-5
Applications Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone HZ52
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Full length human recombinant p21 protein
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Bovine, Human, Rat
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.