Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p300/CBP (CREB-Binding Protein) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS586B Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS586B 1 mL Biotin Concentrated 200μg/mL
MS586P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS586PABX 200 μL Unconjugated Concentrated 1mg/mL
MS586P1 500 μL Unconjugated Concentrated 200μg/mL
MS586R7 7 mL Unconjugated Prediluted Prediluted
MS586B1 500 μL Biotin Concentrated 200μg/mL
MS586P 1 mL Unconjugated Concentrated 200μg/mL
MS586R1 1 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS586B Supplier Epredia Supplier No. MS586B

Achieve accurate, reproducible results with the Epredia™ p300/CBP (CREB-Binding Protein) Ab-1, Mouse Monoclonal Antibody.

p300 is a transcriptional adapter protein which is characterized by three cysteine- and histidine-rich regions and its C-terminus specifically binds the adenovirus E1A protein. p300, and p300-associated proteins, are components of TATA-binding protein (TBP) complexes. Protein kinase A mediated CREB phosphorylation results in the binding of CREB to a 265kDa nuclear protein designated CBP (for CREB-binding protein). CBP and p300 are homologous to each other.

Host Species: Mouse

Clone: NM11

Isotype: IgG2b/κ

Species Reactivity: Human, Monkey, Mink, Mouse, and Rat. Others not known.

Epitope: aa 2071-2091

Immunogen: E1A affinity purified p300 protein

Molecular Weight: 300kDa

Positive Control: Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen p300/CBP (CREB-Binding Protein) Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone NM11
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen E1A affinity purified p300 protein
Quantity 1 mL
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Target Species Human, Mink, Mouse, Non-human Primate, Rat
Form Concentrated
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.