Learn More
Description
Host Species: Mouse
Clone: NM11
Isotype: IgG2b/κ
Species Reactivity: Human, Monkey, Mink, Mouse, and Rat. Others not known.
Epitope: aa 2071-2091
Immunogen: E1A affinity purified p300 protein
Molecular Weight: 300kDa
Positive Control: Tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | p300/CBP (CREB-Binding Protein) Ab-1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | NM11 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | E1A affinity purified p300 protein |
| Quantity | 200 μL |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.