Learn More
Description
Host Species: Rabbit
Clone: 13H4
Isotype: IgG
Species Reactivity: Human. Others not tested.
Immunogen: Human p504s/AMACR polypeptide
Molecular Weight: ∽45 kDa
Positive Control: Prostate carcinoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | p504s/AMACR |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 13H4 |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Human p504s/AMACR polypeptide |
| Quantity | 7 mL |
| Regulatory Status | ASR |
| Research Discipline | Cancer and Tumor Biology |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.