Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p53 Ab-5 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS186PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
12 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Ready-to-Use
Concentration:
1 mg/mL
200 μg/mL
Prediluted
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS186PABX 200 μL Unconjugated Concentrated 1 mg/mL
MS186B1 500 μL Biotin Concentrated 200 μg/mL
MS186B 1 mL Biotin Concentrated 200 μg/mL
MS186P1 500 μL Unconjugated Concentrated 200 μg/mL
MS186P 1 mL Unconjugated Concentrated 200 μg/mL
MS186RQ 12 mL Unconjugated Ready-to-Use Prediluted
MS186B0 100 μL Biotin Concentrated 200 μg/mL
MS186P1ABX 100 μL Unconjugated Concentrated 1 mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS186PABX Supplier Epredia Supplier No. MS186PABX

Achieve accurate, reproducible results with the Epredia™ p53 Ab-5, Mouse Monoclonal Antibody.

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Host Species: Mouse

Clone: DO-7

Isotype: IgG2b/κ

Species Reactivity: Human, Monkey, Cow. Others not known.

Epitope: aa 37-45

Immunogen: Recombinant human wild-type p53 protein expressed in E. coli

Molecular Weight: 53kDa

Positive Control: SKBR-3 or MDA-231 cells, Colon CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)
  • Flow Cytometry

Specifications

Antigen p53 Ab-5
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone DO-7
Concentration 1 mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant human wild-type p53 protein expressed in E. coli
Quantity 200 μL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Bovine, Human, Non-human Primate
Form Concentrated
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.