Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p53 Ab-6 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS187PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS187PABX 200 μL 1mg/mL
MS187P1ABX 100 μL 1mg/mL
MS187P1 500 μL 200μg/mL
MS187P 1 mL 200μg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS187PABX Supplier Epredia Supplier No. MS187PABX

Achieve accurate, reproducible results with the Epredia™ p53 Ab-6, Mouse Monoclonal Antibody.

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Host Species: Mouse

Clone: DO-1

Isotype: IgG2a

Species Reactivity: Human, Monkey and Cow. Reacts weakly with Mouse and Rat. Others not known.

Epitope: aa 11-25

Immunogen: Recombinant human wild-type p53 protein expressed in E. coli

Molecular Weight: 53kDa

Positive Control: SKBR-3 cells or MDA-231 cells, Colon CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry
  • Gel Supershift Assay

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen p53 Ab-6
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone DO-1
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant human wild-type p53 protein expressed in E. coli
Quantity 200 μL
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Target Species Bovine, Human, Non-human Primate
Form Concentrated
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.