Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ p53 Rabbit Monoclonal Antibody (Clone SP5)

Catalog No. RM9105R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM9105R7 7 mL Prediluted
RM9105S0 100 μL Concentrated
RM9105S1 500 μL Concentrated
RM9105RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RM9105R7 Supplier Epredia™ Supplier No. RM9105R7

Employ part of a comprehensive antibody portfolio with the Epredia™ p53, Rabbit Monoclonal Antibody (Clone SP5). Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Host Species: Rabbit

Clone: SP5

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Recombinant human full-length wild type p53 protein

Molecular Weight: 53kDa

Positive Control: MDA-231 cells, Colon CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Specifications

Antigen p53
Applications Western Blot
Classification Monoclonal
Clone SP5
Conjugate Unconjugated
Host Species Rabbit
Immunogen Recombinant human full-length wild type p53 protein
Quantity 7 mL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.