Learn More
Description
Host Species: Rabbit
Clone: Y5
Species Reactivity: Human.Others not known.
Immunogen: A synthetic peptide derived from N- terminal residues of human p53
Molecular Weight: 53kDa
Positive Control: Colon or Breast CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | p53 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | Y5 |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide derived from N- terminal residues of human p53 |
| Quantity | 100 μL |
| Regulatory Status | IVD |
| Research Discipline | Cancer and Tumor Biology |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.