Learn More
Description
Host Species: Mouse
Clone: AE1/AE3
Isotype: IgG1 + IgG1
Species Reactivity: Human, Monkey, Cow, Rabbit, Mouse, Rat and Chicken. Others not known.
Immunogen: Human epidermal keratin
Positive Control: Skin, Lung CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Specifications
Specifications
| Antigen | Cytokeratin, Pan Ab-1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | AE1/AE3 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human epidermal keratin |
| Quantity | 100 μL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.