Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pan-Cytokeratin Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS356P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS356P0 100 μL Concentrated 200μg/mL
MS356P1ABX 100 μL Concentrated 1mg/mL
MS356PABX 200 μL Concentrated 1mg/mL
MS356P1 500 μL Concentrated 200μg/mL
MS356R1 1 mL Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS356P0 Supplier Epredia Supplier No. MS356P0

Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Epredia™ Cytokeratin, Pan Ab-2, Mouse Monoclonal Antibody.

Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI <5.7) and basic (pI >6.0) subfamilies. Members of the acidic and basic subfamilies are found together in pairs. The composition of keratin pairs varies with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

Host Species: Mouse

Clone: PAN-CK

Isotype: IgGs

Species Reactivity: Human. Others not known.

Epitope: Not determined

Positive Control: Skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Cytokeratin, Pan Ab-2
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PAN-CK
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Human
Form Concentrated
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.