Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Pax-5 Rabbit Monoclonal Antibody

Catalog No. RM9133S Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM9133S 1 mL Concentrated
RM9133S0 100 μL Concentrated
RM9133R1 1 mL Prediluted
RM9133R7 7 mL Prediluted
RM9133RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RM9133S Supplier Epredia™ Supplier No. RM9133S

Take advantage of part of a comprehensive antibody portfolio with the Epredia™ Pax-5, Rabbit Monoclonal Antibody.

PAX-5 is a B-cell-specific activator protein (BSAP). In the early stages of B cell developmemt, PAX-5 influences the expression of several B-cell-specific genes, such as CD19 and CD20. PAX-5 is expressed primarily in pro-, pre-, and mature B cells, but not in plasma cells. PAX-5 is expressed in most B-cell malignancies (pre-B and mature B cell lymphomas/leukemias). Over 90% of cases are positive for PAX-5 in Hodgkin's lymphoma, lymphoplasmacytic lymphoma, Merkel cell and small cell carcinoma. T-cell lymphomas, myeloma/plasmacytoma and carcinoid tumors are negative for PAX-5.

Host Species: Rabbit

Clone: SP34

Isotype: IgG

Species Reactivity: Human. Others not tested.

Immunogen: Synthetic peptide derived from the C-terminus of human PAX-5 protein

Molecular Weight: 50kDa

Positive Control: Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen PAX-5
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP34
Conjugate Unconjugated
Host Species Rabbit
Immunogen Synthetic peptide derived from the C-terminus of human Pax-5 protein
Quantity 1 mL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.