Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Pax-5 Rabbit Polyclonal Antibody with BSA and azide

Catalog No. RB9406P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB9406P 1 mL Concentrated
RB9406P0 100 μL Concentrated
RB9406P1 500 μL Concentrated
RB9406R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RB9406P Supplier Epredia™ Supplier No. RB9406P
Add to Cart
Add to Cart

Provide accurate, reproducible results with the Epredia™ Pax-5, Rabbit Polyclonal Antibody.

PAX-5 is a B cell specific activator protein (BSAP). In the early stages of B cell developmemt, PAX-5 influences the expression of several B-cell-specific genes, such as CD19 and CD20. PAX-5 is expressed primarily inpro-, pre-, and mature B cells, but not in plasma cells. Interestingly, PAX-5 mRNA is transiently detected in the mesencephalon and spinal cord during embryogenesis. Expression then shifts to the fetal liver and correlates with the onset of B lymphopoiesis. This indicates that PAX-5 is important in B cell development, but may also have a role in proper neuronal development.

Host Species: Rabbit

Species Reactivity: Human, Mouse, Rat. Others not tested.

Epitope: C-terminal

Immunogen: Synthetic peptide derived from C-terminus of human PAX-5 protein

Molecular Weight: 50kDa

Positive Control: Tonsil, Raji cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Specifications

Antigen PAX-5
Applications Western Blot
Classification Polyclonal
Clone Polyclonal
Concentration 200µg/mL
Conjugate Unconjugated
Host Species Rabbit
Immunogen Synthetic peptide derived from C-terminus of human Pax-5 protein
Quantity 1 mL
Regulatory Status IVD
Research Discipline B-Cell
Primary or Secondary Primary
Target Species Human, Mouse
Form Concentrated
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.