Learn More
Epredia™ Lab Vision™ Pax-5 Rabbit Polyclonal Antibody with BSA and azide
Provide accurate, reproducible results with the Epredia™ Pax-5, Rabbit Polyclonal Antibody.
Supplier: Epredia™ RB9406P1
Description
PAX-5 is a B cell specific activator protein (BSAP). In the early stages of B cell developmemt, PAX-5 influences the expression of several B-cell-specific genes, such as CD19 and CD20. PAX-5 is expressed primarily inpro-, pre-, and mature B cells, but not in plasma cells. Interestingly, PAX-5 mRNA is transiently detected in the mesencephalon and spinal cord during embryogenesis. Expression then shifts to the fetal liver and correlates with the onset of B lymphopoiesis. This indicates that PAX-5 is important in B cell development, but may also have a role in proper neuronal development.Host Species: Rabbit
Species Reactivity: Human, Mouse, Rat. Others not tested.
Epitope: C-terminal
Immunogen: Synthetic peptide derived from C-terminus of human PAX-5 protein
Molecular Weight: 50kDa
Positive Control: Tonsil, Raji cells
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifications
PAX-5 | |
Polyclonal | |
Unconjugated | |
Synthetic peptide derived from C-terminus of human Pax-5 protein | |
IVD | |
Primary | |
Concentrated |
Western Blot | |
200µg/mL | |
Rabbit | |
500 μL | |
B-Cell | |
Human, Mouse |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.