Learn More
Epredia™ Lab Vision™ PCNA (Proliferating Cell Nuclear Antigen) Ab-1 Mouse Monoclonal Antibody, 20μg/mL, biotinylated, BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: PC10
Isotype: IgG2a/κ
Species Reactivity: Human, Mouse, Rat, Insect, and Yeast. Others not known.
Epitope: Not determined
Immunogen: Recombinant rat PCNA protein
Molecular Weight: 36kDa
Positive Control: Raji cells; Tonsil, lymph node, or small intestine
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Native and denatured)
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | PCNA (Proliferating Cell Nuclear Antigen) Ab-1 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | PC10 |
| Concentration | 20μg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Immunogen | Recombinant rat PCNA protein |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.