Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ PCNA (Proliferating Cell Nuclear Antigen) Ab-1 Mouse Monoclonal Antibody, 20μg/mL, biotinylated, BSA and azide

Catalog No. MS106P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
20μg/mL
Prediluted
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS106P1 500 μL Unconjugated Concentrated 20μg/mL
MS106B1 500 μL Biotin Concentrated 20μg/mL
MS106P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS106P 1 mL Unconjugated Concentrated 20μg/mL
MS106R7 7 mL Unconjugated Prediluted Prediluted
MS106B 1 mL Biotin Concentrated 20μg/mL
MS106PABX 200 μL Unconjugated Concentrated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS106P1 Supplier Epredia™ Supplier No. MS106P1
Add to Cart
Add to Cart

Provides accurate, reproducible results with the Epredia™ PCNA (Proliferating Cell Nuclear Antigen) Ab-1, Mouse Monoclonal Antibody.

Expression of proliferating cell nuclear antigen (PCNA) or cyclin or polymerase δ auxiliary protein is elevated in the nucleus during late G1 phase immediately before the onset of DNA synthesis, becoming maximal during S-phase and declining during G2 and M phases. Its level correlates directly with rates of cellular proliferation and DNA synthesis. PCNA/cyclin may act as an auxiliary protein of DNA polymerase-δ to play a fundamental role in the initiation of cell proliferation

Host Species: Mouse

Clone: PC10

Isotype: IgG2a/κ

Species Reactivity: Human, Mouse, Rat, Insect, and Yeast. Others not known.

Epitope: Not determined

Immunogen: Recombinant rat PCNA protein

Molecular Weight: 36kDa

Positive Control: Raji cells; Tonsil, lymph node, or small intestine

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen PCNA (Proliferating Cell Nuclear Antigen) Ab-1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone PC10
Concentration 20μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant rat PCNA protein
Quantity 500 μL
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Target Species Human, Insect, Mouse, Rat, Yeast
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.