Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGFRα Rabbit Polyclonal Antibody, Epredia™

Catalog No. RB9027P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB9027P0 100 μL Concentrated
RB9027P1 500 μL Concentrated
RB9027P 1 mL Concentrated
RB9027R1 1 mL Prediluted
RB9027R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RB9027P0 Supplier Epredia Supplier No. RB9027P0

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Epredia™ PDGFRα, Rabbit Polyclonal Antibody.

PDGFR (Platelet Derived Growth Factor Receptor) exhibits two different isoforms, alpha; and beta; coded by two different genes. PDGF A receptors are expressed in oligodendrocyte progenitor cells and PDGF B receptors are expressed on neurons. This antibody is specific for PDGFR, alpha; and does not react with PDGFR, beta;.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide derived from C-terminal of human PDGFRα

Molecular Weight: 190kDa

Positive Control: Skin and ovarian CA

Cellular Localization: Cytoplasmic and cell membrane This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen PDGFR, alpha
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 200µg/mL
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide derived from C-terminal of human PDGFR alpha
Quantity 100 μL
Regulatory Status IVD
Research Discipline Growth Factor
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.