Learn More
Description
Host Species: Mouse
Clone: DCS-280
Isotype: IgG2a/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Human recombinant full-length Pds1 protein
Molecular Weight: 42kDa
Positive Control: HeLa cells, Tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting (Not verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Pds1 Ab-1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | DCS-280 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human recombinant full length Pds1 protein |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.