Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pds1 Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1511R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1511R7 7 mL Prediluted Prediluted
MS1511P0 100 μL Concentrated 200μg/mL
MS1511P1ABX 100 μL Concentrated 1mg/mL
MS1511PABX 200 μL Concentrated 1mg/mL
MS1511P1 500 μL Concentrated 200μg/mL
MS1511P 1 mL Concentrated 200μg/mL
MS1511R1 1 mL Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS1511R7 Supplier Epredia Supplier No. MS1511R7

Provide accurate, reproducible results with the Epredia™ Pds1 Ab-1, Mouse Monoclonal Antibody.

Pds1 is an anaphase inhibitor and plays important role in DNA damage and spindle check point pathways. Pds1 inhibits sister chromatid separation by binding and inhibiting Esp1, cystein protease that causes cleavage of the cohesin Scc1 that binds the sister chromatids together. Pds1 is responsible for targeting of Esp1 to nucleus and its binding to the spindle. Degradation of Pds1 occurs shortly before anaphase, which liberates Esp1 and is a prerequisite for anaphase entry. Pds1 is targeted for degradation by ubiquitination mediated by cyclosome/anaphase-promoting complex (APC) functioning as a ubiquitin ligase. In response to DNA damage Chk1 phosphorylates Pds1 to stabilize it against the APC mediated destruction, hence preventing the entry of such a cell into anaphase.

Host Species: Mouse

Clone: DCS-280

Isotype: IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human recombinant full-length Pds1 protein

Molecular Weight: 42kDa

Positive Control: HeLa cells, Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Pds1 Ab-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone DCS-280
Concentration Prediluted
Conjugate Unconjugated
Host Species Mouse
Immunogen Human recombinant full length Pds1 protein
Quantity 7 mL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.