Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Perforin Ab-2, Mouse Monoclonal Antibody

Catalog No. MS1834R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1834R1 1 mL Prediluted
MS1834S0 100 μL Concentrated
MS1834S1 500 μL Concentrated
MS1834R7 7 mL Prediluted
MS1834S 1 mL Concentrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1834R1 Supplier Epredia™ Supplier No. MS1834R1

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states Epredia™ Perforin Ab-2, Mouse Monoclonal Antibody.

Perforin is a potent cytolytic pore-forming protein. It is a specific marker of functionally active cytotoxic T-lymphocytes (CTLs) and natural killer (NK) cells. Perforin is a key effector mechanisms in T cell-mediated cytotoxicity. It mediates cytolysis of target cells by membrane damage and apoptosis.

Host Species: Mouse

Clone: 5B10

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: C-terminal

Immunogen: Recombinant protein corresponding to C-terminal region of human perforin

Molecular Weight: 70kDa

Positive Control: Lymphoma

Cellular Localization: Granular and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

General References:

1. Lichtenheld M G, et al. (1988) Nature, 335:448-451.

2. Hameed A, et. al. American Journal of Pathology, 1992, 140(5):1025-30.

Specifications

Antigen Perforin Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5B10
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein corresponding to C-terminal region of human perforin
Quantity 1 mL
Regulatory Status IVD
Research Discipline T-Cell
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.