Learn More
Description
Host Species: Mouse
Clone: 5B10
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: C-terminal
Immunogen: Recombinant protein corresponding to C-terminal region of human perforin
Molecular Weight: 70kDa
Positive Control: Lymphoma
Cellular Localization: Granular and cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Perforin Ab-2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 5B10 |
| Conjugate | Unconjugated |
| Dilution | 1:10 for 60 min at RT |
| Host Species | Mouse |
| Immunogen | Recombinant protein corresponding to C-terminal region of human perforin |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.