Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Phosphate Buffered Saline (25X)

Catalog No. ta125pb Shop All Epredia Products
Click to view available options
Quantity:
125 mL

Dilute Epredia™ Lab Vision™ Phosphate Buffered Saline (25X) before use to make stock solution, which acts as an ideal wash buffer for immunolabeling techniques.

  • For in vitro diagnostic use

Storage Conditions: Store at room temperature. Stable for up to two years. Use diluted solution within one week.

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Concentration 25 X
For Use With (Application) Healthcare
Quantity 125 mL
Solution Type Phosphate Buffered Saline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.