Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Placental Alk. Phosphatase (PLAP) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS208B1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
MS208B1 500 μL Biotin 200μg/mL
MS208B 1 mL Biotin 200μg/mL
MS208P0 100 μL Unconjugated 200μg/mL
MS208P 1 mL Unconjugated 200μg/mL
MS208P1ABX 100 μL Unconjugated 1mg/mL
MS208B0 100 μL Biotin 200μg/mL
MS208P1 500 μL Unconjugated 200μg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS208B1 Supplier Epredia Supplier No. MS208B1

Provide accurate, reproducible results with the Epredia™ Placental Alk. Phosphatase (PLAP) Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: 8B6.18

Isotype: IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: HEp-2 cells expressing Placental Alkaline Phosphatase (PLAP)

Molecular Weight: 60-70kDa

Positive Control: Placenta

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • ELISA

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Placental Alk. Phosphatase (PLAP) Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8B6.18
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen HEp-2 cells expressing Placental Alkaline Phosphatase (PLAP)
Purification Method Placenta
Quantity 500 μL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.