Learn More
Placental Alk. Phosphatase (PLAP) Ab-1 Mouse Monoclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: 8B6.18
Isotype: IgG2a/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: HEp-2 cells expressing Placental Alkaline Phosphatase (PLAP)
Molecular Weight: 60-70kDa
Positive Control: Placenta
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- ELISA
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Placental Alk. Phosphatase (PLAP) Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 8B6.18 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | HEp-2 cells expressing Placental Alkaline Phosphatase (PLAP) |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.