Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Placental Alk. Phosphatase (PLAP) Ab-4, Mouse Monoclonal Antibody

Catalog No. MS734S Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS734S 1 mL Concentrated
MS734S1 500 μL Concentrated
MS734S0 100 μL Concentrated
MS734R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS734S Supplier Epredia™ Supplier No. MS734S
Add to Cart
Add to Cart

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Epredia™ Placental Alk. Phosphatase (PLAP) Ab-4, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: 8A9

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified human Placental Alkaline Phosphatase (PLAP)

Molecular Weight: 60-70kDa

Positive Control: Placenta or seminoma

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Placental Alk. Phosphatase (PLAP) Ab-4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8A9
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified human Placental Alkaline Phosphatase (PLAP)
Quantity 1 mL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.