Learn More
Description
Host Species: Mouse
Clone: hPRa 6
Isotype: IgG2b
Species Reactivity: Human, Cow, Rabbit, and Chicken. Does not react with monkey. Others not known.
Epitope: Not determined
Immunogen: Partially purified PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice
Molecular Weight: 116kDa (triplet, B-form)
Positive Control: T47D cells
Cellular Localization: Nuclear Ab-6 preferentially reacts with B-form of progesterone receptor.
Recommended for:
- Western Blotting (Not optimum)
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Sedimentation in Sucrose Gradient
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Progesterone Receptor Ab-6 |
| Applications | Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | hPRa 6 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Partially purified PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.