Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Progesterone Receptor Ab-6 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS196P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS196P1 500 μL 200μg/mL
MS196PABX 200 μL 1mg/mL
MS196P0 100 μL 200μg/mL
MS196P 1 mL 200μg/mL
MS196P1ABX 100 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS196P1 Supplier Epredia Supplier No. MS196P1

Provide accurate, reproducible results with the Epredia™ Progesterone Receptor Ab-6, Mouse Monoclonal Antibody.

Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.

Host Species: Mouse

Clone: hPRa 6

Isotype: IgG2b

Species Reactivity: Human, Cow, Rabbit, and Chicken. Does not react with monkey. Others not known.

Epitope: Not determined

Immunogen: Partially purified PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice

Molecular Weight: 116kDa (triplet, B-form)

Positive Control: T47D cells

Cellular Localization: Nuclear Ab-6 preferentially reacts with B-form of progesterone receptor.

Recommended for:

  • Western Blotting (Not optimum)
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Sedimentation in Sucrose Gradient

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Progesterone Receptor Ab-6
Applications Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone hPRa 6
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Partially purified PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice
Quantity 500 μL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Bovine, Chicken, Human, Rabbit
Form Concentrated
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.