Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Progesterone Receptor Rabbit Monoclonal Antibody

Catalog No. RM9102R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM9102R1 1 mL Prediluted
RM9102S0 100 μL Concentrated
RM9102S1 500 μL Concentrated
RM9102S 1 mL Concentrated
RM9102R7 7 mL Prediluted
RM9102RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. RM9102R1 Supplier Epredia™ Supplier No. RM9102R1
This item is not returnable. View return policy

Progesterone Receptor, Rabbit Monoclonal Antibody is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

The progesterone receptor (PgR) is an estrogen-regulated protein. It has been proposed that expression of PgR determination indicates a responsive estrogen receptor (ER) pathway, and therefore, may predict likely response to endocrine therapy in human breast cancer. A number of studies have shown that PgR determination provides supplementary information to ER, both in predicting response to endocrine therapy and estimating survival. PgR has proved superior to ER as a prognostic indicator in some studies.

Host Species: Rabbit

Clone: SP2

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: 412-526aa

Immunogen: Recombinant protein encoding human progesterone receptor 412-526aa

Molecular Weight: 116 + 81kDa

Positive Control: Breast CA, T47D cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting


Specifications

Antigen Progesterone Receptor (SP2)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone SP2
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Rabbit
Immunogen Recombinant protein encoding human progesterone receptor 412-526 aa
Quantity 1 mL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.