Learn More
Description
Host Species: Rabbit
Clone: SP2
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: 412-526aa
Immunogen: Recombinant protein encoding human progesterone receptor 412-526aa
Molecular Weight: 116 + 81kDa
Positive Control: Breast CA, T47D cells
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifications
Specifications
| Antigen | Progesterone Receptor (SP2) |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | SP2 |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encoding human progesterone receptor 412-526 aa |
| Quantity | 100 μL |
| Regulatory Status | IVD |
| Research Discipline | Cancer and Tumor Biology |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.