Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prohibitin (Mitochondrial Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS261P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS261P0 100 μL Unconjugated Concentrated 200μg/mL
MS261P1 500 μL Unconjugated Concentrated 200μg/mL
MS261B0 100 μL Biotin Concentrated 200μg/mL
MS261B1 500 μL Biotin Concentrated 200μg/mL
MS261B 1 mL Biotin Concentrated 200μg/mL
MS261P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS261PABX 200 μL Unconjugated Concentrated 1mg/mL
MS261P 1 mL Unconjugated Concentrated 200μg/mL
MS261R1 1 mL Unconjugated Prediluted Prediluted
MS261R7 7 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. MS261P0 Supplier Epredia Supplier No. MS261P0

Provide accurate, reproducible results with the Epredia™ Prohibitin (Mitochondrial Marker) Ab-1, Mouse Monoclonal Antibody.

Prohibitin is an evolutionarily conserved protein located in the inner membrane of mitochondria. Prohibitin appears to be a very reliable marker of mitochondria. Prohibitin shows antiproliferative activity and has been proposed to play a role in normal cell cycle regulation, replicative senescence, cellular immortalization, and tumor suppression.

Host Species: Mouse

Clone: II-14-10

Isotype: IgG1

Species Reactivity: Human, Pig, Rat, Mouse, and Chicken. Others not known.

Epitope: Not determined

Immunogen: Purified recombinant rat prohibitin protein

Molecular Weight: 30kDa

Positive Control: MAD109 or PC12 cells, Heart

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Prohibitin (Mitochondrial Marker) Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone II-14-10
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified recombinant rat prohibitin protein
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Target Species Chicken, Human, Mouse, Pig, Rat
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.