Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prolactin Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1362P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1362P1 500 μL Concentrated 200μg/mL
MS1362P1ABX 100 μL Concentrated 1mg/mL
MS1362PABX 200 μL Concentrated 1mg/mL
MS1362P0 100 μL Concentrated 200μg/mL
MS1362R1 1 mL Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1362P1 Supplier Epredia Supplier No. MS1362P1
Add to Cart
Add to Cart

Provide accurate, reproducible results with the Epredia™ Prolactin Ab-2, Mouse Monoclonal Antibody.

Prolactin is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. Prolactin producing cells make up approximately 20% of the pituitary.

Host Species: Mouse

Clone: PRL02

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human prolactin

Molecular Weight: 23.5kDa

Positive Control: Pituitary gland

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • ELISA

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Prolactin Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PRL02
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human prolactin
Quantity 500 μL
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.