Learn More
Epredia™ Lab Vision™ Prolactin Mouse Monoclonal Antibody with BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: SPM108
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Immunogen: Purified human prolactin
Molecular Weight: 23.5kDa
Positive Control: Pituitary gland
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Prolactin |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | SPM108 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Purified human prolactin |
| Quantity | 100 μL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.