Learn More
PTEN/MMAC1 Ab-4 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ PTEN/MMAC1 Ab-4, Mouse Monoclonal Antibody.
Supplier: Epredia MS1601S1
Description
PTEN/MMAC1, a tumor suppressor, has been implicated in a large number of human tumors and is conserved from humans to worms. Characterization of PTEN protein showed that it is a phosphatase that acts on proteins and on 3-phosphorylated phosphoinositides, and can therefore modulate signal-transduction pathways that involve lipid second messengers. Recent results indicate that at least part of its role is to regulate the activity of the serine/threonine kinase AKT/PKB, and thus influence cell survival signaling.Host Species: Mouse
Clone: 17.A
Isotype: IgM/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant human PTEN protein
Molecular Weight: 60kDa
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry
Note:
USA: RUO; Int'l: RUO
Specifications
PTEN/MMAC1 Ab-4 | |
Monoclonal | |
Unconjugated | |
Recombinant human PTEN protein | |
RUO | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
17.A | |
Mouse | |
500 μL | |
Cancer and Tumor Biology | |
Human | |
IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.