Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Rhodopsin (Opsin) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Catalog No. MS1233B1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1233B1 500 μL Concentrated 200μg/mL
MS1233P1 500 μL Concentrated 200μg/mL
MS1233B0 100 μL Concentrated 200μg/mL
MS1233P0 100 μL Concentrated 200μg/mL
MS1233P1ABX 100 μL Concentrated 1mg/mL
MS1233PABX 200 μL Concentrated 1mg/mL
MS1233B 1 mL Concentrated 200μg/mL
MS1233P 1 mL Concentrated 200μg/mL
MS1233R7 7 mL Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. MS1233B1 Supplier Epredia™ Supplier No. MS1233B1

Provide accurate, reproducible results with the Epredia™ Rhodopsin (Opsin) Ab-1, Mouse Monoclonal Antibody.

Rhodopsin is a glycoprotein which is a visual pigment in mammals, birds, amphibians, and reptiles. The rhodopsin protein is present on the photoreceptor cells in the retina.

Host Species: Mouse

Clone: RET-P1

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Membrane preparation from adult rat retina

Molecular Weight: 39kDa

Positive Control: Brain or retina

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Rhodopsin (Opsin) Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone RET-P1
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen Membrane preparation from adult rat retina
Quantity 500 μL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.